Tisk…

Methyl cinnamate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate

M-Clarity™ quality level = MQ300

W269816-100G-K W269816-1KG-K
SIALW269816-100G-KEA 6150 CZK
SIALW269816-100G-K SIALW269816-1KG-K
Methyl cinnamate, Sigma-Aldrich®
Methyl cinnamate
Vzorec: C₁₀H₁₀O₂
MW: 162,19 g/mol
MDL Number: MFCD00008458
CAS číslo: 103-26-4
EINECS: 203-093-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR