Tisk…

Methyl trans-cinnamate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl (E)-cinnamate, trans-Cinnamic acid methyl ester

M-Clarity™ quality level = MQ300

W269808-10KG-K W269808-1KG-K W269808-25KG-K
SIALW269808-10KG-KEA 10920 CZK
SIALW269808-10KG-K SIALW269808-1KG-K SIALW269808-25KG-K
Methyl trans-cinnamate, Sigma-Aldrich®
Methyl trans-cinnamate
Vzorec: C₁₀H₁₀O₂
MW: 162,19 g/mol
Bod varu: 260…262 °C (1013 hPa)
Bod tání: 34 °C
Hustota: 1,08 g/cm³ (20 °C)
MDL Number: MFCD00008458
CAS číslo: 1754-62-7
EINECS: 203-093-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR