Ethylcinnamát ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A12906.0E
A12906.22
A12906.36
A12906.0EEA
13630
CZK
A12906.0E
A12906.22
A12906.36
Ethylcinnamát ≥98%
Ethylcinnamát
Purity: 98+%
CAS No.: 103-36-6
Molecular Formula: C11H12O2
Formula Weight: 176.22
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 1.049
Refractive Index: 1.5590
Appearance: Colourless
Melting Point: 7°
Boiling Point: 270-272°
Beilstein Reference: 1238804
Merck Reference: 14,2299
MDL No.: MFCD00009189
Vzorec:
C₁₁H₁₂O₂ Bod varu: 270…272 °C Bod tání: 7 °C Hustota: 1,049 Bod vzplanutí: 135 °C (275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009189 CAS číslo: 103-36-6 EINECS: 203-104-6 Merck Index: 14,2299 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...