Tisk…

γ-Linolenic acid methyl ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl γ-linolenate, Methyl cis,cis,cis-6,9,12-octadecatrienoate, 6Z,9Z,12Z-Octadecatrienoic acid methyl ester, Methyl (6Z,9Z,12Z)-6,9,12-octadecatrienoate, (6Z,9Z,12Z)-Methyl octadeca-6,9,12-trienoate, cis,cis,cis-6,9,12-Octadecatrienoic acid methyl ester

M-Clarity™ quality level = MQ100

L6503-100MG L6503-1G L6503-500MG
SIALL6503-100MGEA 6270 CZK
SIALL6503-100MG SIALL6503-1G SIALL6503-500MG
γ-Linolenic acid methyl ester, Sigma-Aldrich®
γ-Linolenic acid methyl ester
Vzorec: C₁₉H₃₂O₂
MW: 292,46 g/mol
MDL Number: MFCD00010455
CAS číslo: 16326-32-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR