γ-Linolenic acid methyl ester
Dodavatel: Cayman Chemical
Synonyms:
Methyl γ-linolenate, Methyl cis,cis,cis-6,9,12-octadecatrienoate, 6Z,9Z,12Z-Octadecatrienoic acid methyl ester, Methyl (6Z,9Z,12Z)-6,9,12-octadecatrienoate, (6Z,9Z,12Z)-Methyl octadeca-6,9,12-trienoate, cis,cis,cis-6,9,12-Octadecatrienoic acid methyl ester
10006579-100
10006579-50
10006579-250
10006579-1
CAYM10006579-100EA
2490
CZK
CAYM10006579-100
CAYM10006579-50
CAYM10006579-250
CAYM10006579-1
γ-Linolenic acid methyl ester
γ-Linolenic acid methyl ester
Vzorec:
C₁₉H₃₂O₂ MW: 292.46 |
MDL Number:
MFCD00010455 CAS číslo: 16326-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...