Tisk…
Toluen-α-sulfonylfluorid, Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride
M-Clarity™ quality level = MQ300
P7626-5G P7626-250MG P7626-100G P7626-1G P7626-25G
SIALP7626-5GEA 3390 CZK
SIALP7626-5G SIALP7626-250MG SIALP7626-100G SIALP7626-1G SIALP7626-25G
Toluen-α-sulfonylfluorid, Sigma-Aldrich®
Toluen-α-sulfonylfluorid
Vzorec: C₇H₇FO₂S
MW: 174.2
Bod varu: 80…85 °C (0.3 mmHg)
Bod tání: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS číslo: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8 III