Tisk…

Toluene-α-sulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Benzylsulphonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride

M-Clarity™ quality level = MQ200

159719-25G 159719-5G
SIAL159719-25GEA 4580 CZK
SIAL159719-25G SIAL159719-5G
Toluene-α-sulphonyl chloride, Sigma-Aldrich®
Toluene-α-sulphonyl chloride
Vzorec: C₇H₇ClO₂S
MW: 190,65 g/mol
Bod varu: 120 °C (10 mmHg)
Bod tání: 90…93 °C
MDL Number: MFCD00007455
CAS číslo: 1939-99-7
EINECS: 217-717-1
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR