Toluene-α-sulphonyl chloride ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Benzylsulphonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride
A10124.06
A10124.14
A10124.22
A10124.06EA
930
CZK
A10124.06
A10124.14
A10124.22
Toluene-α-sulphonyl chloride ≥99%
Toluene-α-sulphonyl chloride
Purity: 99%
CAS No.: 1939-99-7
Molecular Formula: C6H5CH2SO2Cl
Formula Weight: 190.65
Melting Point: 89-94°
Boiling Point: 120°/10mm
UN No.: 3096
MDL No.: MFCD00007455
Vzorec:
C₇H₇ClO₂S Bod varu: 120 °C (10 mmHg) Bod tání: 89…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007455 CAS číslo: 1939-99-7 EINECS: 217-717-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...