Tisk…

Toluene-α-sulphonyl chloride 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Benzylsulphonyl chloride, α-Toluenesulphonyl chloride, Phenylmethylsulphonyl chloride, Phenylmethanesulphonyl chloride

170490050 170490250
ACRO170490050EA 1340 CZK
ACRO170490050 ACRO170490250
Toluene-α-sulphonyl chloride 98%
Toluene-α-sulphonyl chloride
Vzorec: C₇H₇ClO₂S
Bod varu: 120 °C (10 mmHg)
Bod tání: 90…94 °C
MDL Number: MFCD00007455
CAS číslo: 1939-99-7
EINECS: 217-717-1
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
Titration Argentometric ≥97.5 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR