Tisk…

2-Fenylethylacetát, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Phenylethyl Acetate, 2-Phenethyl acetate, Phenethyl acetate

M-Clarity™ quality level = MQ100

290580-250ML 290580-5ML
SIAL290580-250MLEA 1230 CZK
SIAL290580-250ML SIAL290580-5ML
2-Fenylethylacetát, Sigma-Aldrich®
2-Fenylethylacetát
Vzorec: C₁₀H₁₂O₂
MW: 164,2 g/mol
Bod varu: 233 °C (1013 hPa)
Bod tání: –31 °C
Hustota: 1,0286 g/cm³ (20 °C)
MDL Number: MFCD00008720
CAS číslo: 103-45-7
EINECS: 203-113-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR