Benzyl acetoacetate ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A14915.14
A14915.18
A14915.22
A14915.30
A14915.14EA
1100
CZK
A14915.14
A14915.18
A14915.22
A14915.30
Benzyl acetoacetate ≥97%
Benzyl acetoacetate
Purity: 97%
CAS No.: 5396-89-4
Molecular Formula: C11H12O3
Formula Weight: 192.22
Flash Point: 108°(226°F)
Refractive Index: 1.5120
Boiling Point: 276°
Beilstein Reference: 2049615
MDL No.: MFCD00008785
Vzorec:
C₁₁H₁₂O₃ MW: 192,21 g/mol Bod varu: 276 °C Hustota: 1,113 Bod vzplanutí: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008785 CAS číslo: 5396-89-4 EINECS: 226-416-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...