Tisk…
Copper(II)-2-ethylhexanoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 2-Ethylhexanoic acid copper (II) salt, Copper(II) 2-ethylhexanoate, 2-Ethylcaproic acid copper (II) salt, Copper bis(2-ethylhexanoate)
M-Clarity™ quality level = MQ100
337323-25G 337323-5G
SIAL337323-25GEA 5780 CZK
SIAL337323-25G SIAL337323-5G
Copper(II)-2-ethylhexanoate, Sigma-Aldrich®
Copper(II)-2-ethylhexanoate
Vzorec: C₁₆H₃₀CuO₄
MW: 349.96
Bod tání: 252
MDL Number: MFCD00015695
CAS číslo: 149-11-1
EINECS: 205-731-0