Tisk…

2',3'-O-Isopropylideneadenosine ≥98.0% (by HPLC, titration analysis)

Dodavatel: TCI

Synonyms: 2',3'-o-Isopropylideneadenosine

I0702-25G I0702-5G
TCIAI0702-25GEA 6970 CZK
TCIAI0702-25G TCIAI0702-5G
2',3'-O-Isopropylideneadenosine ≥98.0% (by HPLC, titration analysis)
2',3'-O-Isopropylideneadenosine
Vzorec: C₁₃H₁₇N₅O₄
MW: 307,31 g/mol
MDL Number: MFCD00005756
CAS číslo: 362-75-4
EINECS: 206-650-3

Objednat

Výsledky specifikačního testu

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 220.0 to 225.0 deg-C
Specific rotation [a]20/D -100.0 to -110.0 deg (C=1, Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR