Tisk…

Ethyl isobutyrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methylpropionic acid ethyl ester

M-Clarity™ quality level = MQ300

W242810-100G-K W242810-1KG-K
SIALW242810-100G-KEA 5310 CZK
SIALW242810-100G-K SIALW242810-1KG-K
Ethyl isobutyrate, Sigma-Aldrich®
Ethyl isobutyrate
Vzorec: C₆H₁₂O₂
MW: 116,16 g/mol
Bod varu: 112 °C (1013 hPa)
Bod tání: –88 °C
Hustota: 0,87 g/cm³ (20 °C)
MDL Number: MFCD00009165
CAS číslo: 97-62-1
EINECS: 202-595-4
UN: 2385
ADR: 3,II
Merck Index: 13,03847

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR