Tisk…

Ethyl isobutyrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methylpropionic acid ethyl ester

M-Clarity™ quality level = MQ300

W242802-1KG-K W242802-20KG-K W242802-9KG-K
SIALW242802-1KG-KEA 2800 CZK
SIALW242802-1KG-K SIALW242802-20KG-K SIALW242802-9KG-K
Ethyl isobutyrate, Sigma-Aldrich®
Ethyl isobutyrate
Vzorec: C₆H₁₂O₂
MW: 116.16
Bod varu: 112
Bod tání: –88 °C
Hustota: 0.87
MDL Number: MFCD00009165
CAS číslo: 97-62-1
EINECS: 202-595-4
UN: 2385
ADR: 3 II
Merck Index: 13,03847

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR