Ethyl 2-bromoisobutyrate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Ethyl α-bromoisobutyrate, Ethyl 2-bromo-2-methylpropionate, 2-Brom-2-methylpropionic acid ethyl ester
B22785.18
B22785.30
B22785.18EA
1150
CZK
B22785.18
B22785.30
Ethyl 2-bromoisobutyrate ≥98%
Ethyl 2-bromoisobutyrate
Purity: 98+%
CAS No.: 600-00-0
Molecular Formula: C6H11BrO2
Formula Weight: 195.06
Flash Point: 57°(135°F)
Refractive Index: 1.4440
Boiling Point: 162-164°
Beilstein Reference: 1098947
UN No.: 2924
MDL No.: MFCD00000123
Vzorec:
C₆H₁₁BrO₂ MW: 195.06 Bod varu: 162…164 °C Hustota: 1.329 Bod vzplanutí: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000123 CAS číslo: 600-00-0 EINECS: 209-980-6 UN: 2924 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...