Tisk…

3-(tert-Butoxycarbonylamino)-1-propanol ≥96.0% (by GC)

Synonyms: 3-(Boc-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid tert-Butyl Ester, Boc-β-Ala-ol, 3-(BOC-amino)-1-propanol, tert-Butyl N-(3-Hydroxypropyl)carbamate

H0900-25ML H0900-5ML
TCIAH0900-25MLEA 4990 CZK
TCIAH0900-25ML TCIAH0900-5ML
3-(tert-Butoxycarbonylamino)-1-propanol ≥96.0% (by GC)
3-(tert-Butoxycarbonylamino)-1-propanol
Vzorec: C₈H₁₇NO₃
MW: 175,23 g/mol
MDL Number: MFCD00191883
CAS číslo: 58885-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=360c981af42765a58a7acb48453ceee35fae1f52&vid=360c981af42765a58a7acb48453ceee35fae1f52&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR