Tisk…

Itaconic acid 99+%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Methylenesuccinic acid, Methylenesuccinic acid

122810010 122812500 122810051
ACRO122810010EA 1850 CZK
ACRO122810010 ACRO122812500 ACRO122810051
Itaconic acid 99+%
Itaconic acid
Vzorec: C₅H₆O₄
MW: 130,1 g/mol
Bod varu: 381 °C (1013 hPa)
Bod tání: 165…168 °C
Hustota: 1,57 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004260
CAS číslo: 97-65-4
EINECS: 202-599-6
Merck Index: 13,05264

Objednat

Výsledky specifikačního testu

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 165 to 168 °C
Water ≤0.5 %
Sulfated ash ≤0.1 %
Titration with NaOH ≥99.0 %
Sulfate (SO4) ≤300 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR