Tisk…

Dimethyl itaconate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester

M-Clarity™ quality level = MQ100

109533-100G 109533-500G
SIAL109533-100GEA 1650 CZK
SIAL109533-100G SIAL109533-500G
Dimethyl itaconate, Sigma-Aldrich®
Dimethyl itaconate
Vzorec: C₇H₁₀O₄
MW: 158,15 g/mol
Bod varu: 208 °C (1013 hPa)
Bod tání: 38 °C
Hustota: 1,1399 g/cm³ (14.7 °C)
MDL Number: MFCD00008446
CAS číslo: 617-52-7
EINECS: 210-519-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR