Tisk…

1,1,3,3-Tetramethoxypropane, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Malonaldehyde bis(dimethyl acetal)

M-Clarity™ quality level = MQ200

108383-100ML 108383-500ML
SIAL108383-100MLEA 1150 CZK
SIAL108383-100ML SIAL108383-500ML
1,1,3,3-Tetramethoxypropane, Sigma-Aldrich®
1,1,3,3-Tetramethoxypropane
Vzorec: C₇H₁₆O₄
MW: 164,2 g/mol
Bod varu: 183 °C (1013 hPa)
Hustota: 0,99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008488
CAS číslo: 102-52-3
EINECS: 203-037-2
UN: 3271
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR