Dihexyl ether ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
L07076.09
L07076.18
L07076.09EA
1110
CZK
L07076.09
L07076.18
Dihexyl ether ≥98%
Dihexyl ether
Purity: 98%
CAS No.: 112-58-3
Molecular Formula: C12H26O
Formula Weight: 186.34
Flash Point: 78°(172°F)
Refractive Index: 1.4204
Odour: Mild
Melting Point: -43°
Boiling Point: 228-230°
Beilstein Reference: 1738177
MDL No.: MFCD00009525
Vzorec:
C₁₂H₂₆O Bod varu: 228…230 °C Bod tání: –43 °C Hustota: 0,793 Bod vzplanutí: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009525 CAS číslo: 112-58-3 EINECS: 203-987-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...