Dimethylglyoxim, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime
M-Clarity™ quality level = MQ200
162574-100G
162574-500G
SIAL162574-100GEA
4030
CZK
SIAL162574-100G
SIAL162574-500G
Dimethylglyoxim, Sigma-Aldrich®
Dimethylglyoxim
Vzorec:
CH₃C(=NOH)C(=NOH)CH₃ MW: 116,12 g/mol Bod varu: 53 °C (8 mmHg) Bod tání: 240…241 °C Hustota: 1,37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002117 CAS číslo: 95-45-4 EINECS: 202-420-1 Merck Index: 13,03272 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...