Dimethylglyoxim, white powder ACS
Dodavatel: MP Biomedicals
|
Nebezpečí
|
Synonyms:
Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime
0215251880
0215251890
0215251825
ICNA0215251880EA
3220
CZK
ICNA0215251880
ICNA0215251890
ICNA0215251825
Dimethylglyoxim, white powder ACS
Dimethylglyoxim
Soluble in alcohol, ether, pyridine, acetone; practically insoluble in water.
- Insoluble in alcohol: ≤0,05%
Vzorec:
CH₃C(=NOH)C(=NOH)CH₃ Bod varu: 53 °C (8 -rr) Bod tání: ~240 °C Hustota: 1,37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002117 CAS číslo: 95-45-4 EINECS: 202-420-1 UN: 2926 ADR: 4.1,III Merck Index: 13,03272 |
Výsledky specifikačního testu
Identity Test | Passes |
Melting Point | ~240°C |
Suitability for Ni determination | Passes |
Insoluble In Alcohol | ≤0,05% |
Residue on Ignition | ≤0,05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...