Dimethylglyoxim ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime
A12810.0E
A12810.22
A12810.36
A12810.0EEA
22720
CZK
A12810.0E
A12810.22
A12810.36
Dimethylglyoxim ≥99%
Dimethylglyoxim
Purity: 99%
CAS No.: 95-45-4
Molecular Formula: C4H8N2O2
Formula Weight: 116.12
Melting Point: ca 240° dec.
Beilstein Reference: 506731
Merck Reference: 14,3246
MDL No.: MFCD00002117
Vzorec:
CH₃C(=NOH)C(=NOH)CH₃ MW: 116,12 g/mol Bod varu: 53 °C (8 mmHg) Bod tání: ∼ 240 °C (rozkl.) Hustota: 1,37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002117 CAS číslo: 95-45-4 EINECS: 202-420-1 Merck Index: 14,3246 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...