Octadecylamine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Octadecan-1-amine, 1-Octadecylamine, Stearylamine, 1-Aminooctadecane
M-Clarity™ quality level = MQ200
305391-100G
305391-25G
SIAL305391-100GEA
19150
CZK
SIAL305391-100G
SIAL305391-25G
Octadecylamine, Sigma-Aldrich®
Octadecylamine
Vzorec:
H₃C(CH₂)₁₆CH₂NH₂ MW: 269.51 Bod varu: 349 Bod tání: 50…52 °C Hustota: 0.8618 Bod vzplanutí: 110 Storage Temperature: Ambient |
MDL Number:
MFCD00008159 CAS číslo: 124-30-1 EINECS: 204-695-3 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...