2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
BES-Na, BES sodium salt, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid sodium salt, Sodium 2-(bis(2-hydroxyethyl)amino)ethanesulphonate
M-Clarity™ quality level = MQ200
B2891-100G
B2891-25G
B2891-500G
SIALB2891-100GEA
14280
CZK
SIALB2891-100G
SIALB2891-25G
SIALB2891-500G
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt, Sigma-Aldrich®
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt
Vzorec:
C₆H₁₄NNaO₅S MW: 235,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00067425 CAS číslo: 66992-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...