Kyselina 2-[bis(2-hydroxyetyl)amino]ethansulfonová ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
BES, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid
A16092.14
A16092.22
A16092.36
A16092.14EA
1150
CZK
A16092.14
A16092.22
A16092.36
Kyselina 2-[bis(2-hydroxyetyl)amino]ethansulfonová ≥99%
Kyselina 2-[bis(2-hydroxyetyl)amino]ethansulfonová
Purity: 99%
CAS No.: 10191-18-1
Molecular Formula: C6H15NO5S
Formula Weight: 213.25
Melting Point: 153-157°
Solubility: Soluble in water
Beilstein Reference: 1781572
MDL No.: MFCD00007533
Vzorec:
C₆H₁₅NO₅S MW: 213,25 g/mol Bod varu: 405 °C (1013 hPa) Bod tání: 153…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007533 CAS číslo: 10191-18-1 EINECS: 233-465-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...