3-(Diethylamino)-1-propanol 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-(Diethylamino)propan-1-ol, 3-Diethylamino-1-propanol
165410050
165410500
ACRO165410050EA
978
CZK
ACRO165410050
ACRO165410500
3-(Diethylamino)-1-propanol 95%
3-(Diethylamino)-1-propanol
Vzorec:
C₇H₁₇NO MW: 131,22 g/mol Bod varu: 81…83 °C (15 mmHg) Hustota: 0,88 g/cm³ Bod vzplanutí: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002947 CAS číslo: 622-93-5 EINECS: 210-759-1 UN: 2735 ADR: 8,III |
Výsledky specifikačního testu
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4425 to 1.4445 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.003 % |
Aluminium (Al) | ≤0.5 ppm |
Antimony (Sb) | ≤0.02 ppm |
Arsenic (As) | ≤0.05 ppm |
Barium (Ba) | ≤0.1 ppm |
Bismuth (Bi) | ≤0.1 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Lithium (Li) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Mercury (Hg) | ≤0.05 ppm |
Molybdenum (Mo) | ≤0.1 ppm |
Nickel (Ni) | ≤0.02 ppm |
Potassium (K) | ≤0.1 ppm |
Sodium (Na) | ≤1 ppm |
Strontium (Sr) | ≤0.2 ppm |
Zinc (Zn) | ≤0.1 ppm |
Carbonyl (as HCHO) | ≤0.1 % |
Acidity (CH3COOH) | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...