Ethyl iodoacetate ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
B24177.06
B24177.14
B24177.22
B24177.06EA
1040
CZK
B24177.06
B24177.14
B24177.22
Ethyl iodoacetate ≥98%
Ethyl iodoacetate
Purity: 98%
CAS No.: 623-48-3
Molecular Formula: ICH2CO2CH2CH3
Formula Weight: 214
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Refractive Index: 1.503
Boiling Point: 179-180°
Beilstein Reference: 741934
UN No.: 2927
MDL No.: MFCD00001081
Vzorec:
C₄H₇IO₂ MW: 214 g/mol Bod varu: 179…180 °C Hustota: 1,81 Bod vzplanutí: 76 °C (169 °F) |
MDL Number:
MFCD00001081 CAS číslo: 623-48-3 EINECS: 210-796-3 UN: 2927 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...