Tisk…

Sodium trifluoroacetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sodium 2,2,2-trifluoroacetate

M-Clarity™ quality level = MQ100

132101-100G 132101-25G
SIAL132101-100GEA 3950 CZK
SIAL132101-100G SIAL132101-25G
Sodium trifluoroacetate, Sigma-Aldrich®
Sodium trifluoroacetate
Vzorec: C₂F₃NaO₂
MW: 136,01 g/mol
Bod tání: 207…209 °C
MDL Number: MFCD00013217
CAS číslo: 2923-18-4
EINECS: 220-879-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR