Methyl carbamate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Carbamic acid methyl ester
M-Clarity™ quality level = MQ100
246352-100G
246352-500G
SIAL246352-100GEA
1150
CZK
SIAL246352-100G
SIAL246352-500G
Methyl carbamate, Sigma-Aldrich®
Methyl carbamate
Vzorec:
C₂H₅NO₂ MW: 75,07 g/mol Bod varu: 177 °C (1013 hPa) Bod tání: 53…54 °C Hustota: 1,1361 g/cm³ (56 °C) |
MDL Number:
MFCD00007964 CAS číslo: 598-55-0 EINECS: 209-939-2 Merck Index: 12,06115 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...