Tisk…

8-Aminocaprylic acid 99%

Dodavatel: Thermo Fisher Scientific

A17353.06 A17353.03 A17353.14
A17353.06EA 6010 CZK
A17353.06 A17353.03 A17353.14
8-Aminocaprylic acid 99%
8-Aminocaprylic acid

Purity: 99% CAS No.: 1002-57-9 Molecular Formula: NH2(CH2)7CO2H Formula Weight: 159.23 Melting Point: 193-196° MDL No.: MFCD00008245

Vzorec: C₈H₁₇NO₂
Bod varu: 289 °C (1013 hPa)
Bod tání: 193…196 °C
Hustota: 1,221 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008245
CAS číslo: 1002-57-9
EINECS: 213-687-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR