7-Aminoenanthic acid 98%
Dodavatel: Thermo Fisher Scientific
H27519.03
H27519.06
H27519.03EA
4090
CZK
H27519.03
H27519.06
7-Aminoenanthic acid 98%
7-Aminoenanthic acid
Purity: 98%
CAS No.: 929-17-9
Molecular Formula: H2N(CH2)6CO2H
Formula Weight: 145.19
Melting Point: 194-196°
Beilstein Reference: 906887
Vzorec:
C₇H₁₅NO₂ Bod tání: 194…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008242 CAS číslo: 929-17-9 EINECS: 213-197-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...