jodoctan sodný ≥99.5% (by total nitrogen and titration analysis), BioUltra, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Sodium 2-iodoacetate, Iodoacetic Acid, Sodium Salt
57858-25G-F
57858-5G-F
SIAL57858-25G-FEA
12930
CZK
SIAL57858-25G-F
SIAL57858-5G-F
jodoctan sodný ≥99.5% (by total nitrogen and titration analysis), BioUltra, Sigma-Aldrich®
jodoctan sodný
Reagent for the modification of cysteine residues in proteins
- Assay: ≥99.5%
- Solubility: Methanol Soluble (0.1 M at 20 °C), clear and colorless
- Melting Point: 208 °C (dec.)(lit.)
Used in protein sequencing (including by sanger in sequencing bovine insulin) to prevent oxidation of -SH sidechains and to induce animal osteoarthritis model system, which mimics both symptoms and histopathology of the human disease.
Vzorec:
C₂H₂INaO₂ Bod tání: 208…210 °C Bod vzplanutí: 208 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002686 CAS číslo: 305-53-3 EINECS: 206-165-7 UN: 2811 ADR: 6.1,III |
Výsledky specifikačního testu
Insoluble matter | passes filter test |
Water | ≤0.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...