Tisk…

Octan lithný dihydrát, reagent

Dodavatel: Thermo Fisher Scientific

Synonyms: Acetic acid lithium salt

13417.30 13417.A1
13417.30EA 1890 CZK
13417.30 13417.A1
Octan lithný dihydrát, reagent
Octan lithný dihydrát

CAS No.: 6108-17-4 Molecular Formula: LiOOCCH3.2H2O Formula Weight: 102.01 (65.98anhy) Physical Form: Crystals Density: 1.300 Appearance: White Melting Point: 54-57° Solubility: Freely soluble in water and alcohol Beilstein Reference: 3564320 Merck Reference: 14,5521 MDL No.: MFCD00066949

Vzorec: H₃CCOOLi·2H₂O
MW: 102.02
Bod tání: 54…57 °C
Hustota: 1.300
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS číslo: 6108-17-4
EINECS: 208-914-3
Merck Index: 14,5521

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR