Butyl-3-mercaptopropionate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
381454-100ML
381454-500ML
SIAL381454-100MLEA
2420
CZK
SIAL381454-100ML
SIAL381454-500ML
Butyl-3-mercaptopropionate, Sigma-Aldrich®
Butyl-3-mercaptopropionate
Vzorec:
C₇H₁₄O₂S |
CAS číslo:
16215-21-7 EINECS: 240-343-5 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...