Tisk…

2-Butoxyethylacetát ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Butylglycol acetate

L09515.AE L09515.AP L09515.0F
L09515.AEEA 797 CZK
L09515.AE L09515.AP L09515.0F
2-Butoxyethylacetát ≥98%
2-Butoxyethylacetát

Purity: 98% CAS No.: 112-07-2 Molecular Formula: C8H16O3 Formula Weight: 160.21 Flash Point: 71°(159°F) Refractive Index: 1.4130 Odour: Fruit-like Melting Point: -64° Boiling Point: 191-192° Beilstein Reference: 1756960 MDL No.: MFCD00009457

Vzorec: C₈H₁₆O₃
Bod varu: 191…192 °C
Bod tání: –64 °C
Hustota: 0,942
Bod vzplanutí: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009457
CAS číslo: 112-07-2
EINECS: 203-933-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR