Methyl undecanoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Undecanoic acid methyl ester, Undecylic acid methyl ester
M-Clarity™ quality level = MQ200
U0250-5G
U0250-10G
U0250-25G
SIALU0250-5GEA
2530
CZK
SIALU0250-5G
SIALU0250-10G
SIALU0250-25G
Methyl undecanoate, Sigma-Aldrich®
Methyl undecanoate
Vzorec:
C₁₂H₂₄O₂ |
MDL Number:
MFCD00008957 CAS číslo: 1731-86-8 EINECS: 217-053-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...