Tisk…

Methyl undecanoate ≥99%

Dodavatel: Thermo Fisher Scientific

L05939.06 L05939.14
L05939.06EA 1120 CZK
L05939.06 L05939.14
Methyl undecanoate ≥99%
Methyl undecanoate

Purity: 99% CAS No.: 1731-86-8 Molecular Formula: CH3(CH2)9CO2CH3 Flash Point: 109°(228°F) Refractive Index: 1.4290 Melting Point: -10° Boiling Point: 246-248° Beilstein Reference: 1762129 MDL No.: MFCD00008957

Vzorec: C₁₂H₂₄O₂
Bod varu: 246…248 °C
Bod tání: –10 °C
Hustota: 0,871
Bod vzplanutí: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008957
CAS číslo: 1731-86-8
EINECS: 217-053-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR