Stearyl stearate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Stearic acid stearyl ester, Octadecyl stearate, Octadecyl octadecanoate
85775-1G
85775-5G
SIAL85775-1GEA
9290
CZK
SIAL85775-1G
SIAL85775-5G
Stearyl stearate, Sigma-Aldrich®
Stearyl stearate
Vzorec:
C₃₆H₇₂O₂ MW: 536,97 g/mol Bod varu: ∼175 °C (1013 hPa) Bod tání: 64 °C Bod vzplanutí: >160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00056225 CAS číslo: 2778-96-3 EINECS: 220-476-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...