Methyl palmitate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester
M-Clarity™ quality level = MQ300
W509531-1KG
W509531-4KG
W509531-8KG
W509531-SAMPLE
SIALW509531-1KGEA
4410
CZK
SIALW509531-1KG
SIALW509531-4KG
SIALW509531-8KG
SIALW509531-SAMPLE
Methyl palmitate, Sigma-Aldrich®
Methyl palmitate
Vzorec:
C₁₇H₃₄O₂ MW: 270,46 g/mol Bod varu: 417 °C (1013 hPa) Bod tání: 29,9 °C Hustota: 0,8804 g/cm³ (20 °C) |
MDL Number:
MFCD00008994 CAS číslo: 112-39-0 EINECS: 203-966-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...