Tisk…

Methyl palmitate ≥97%

Dodavatel: Thermo Scientific

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

L05509.22 L05509.36
L05509.22EA 1620 CZK
L05509.22 L05509.36
Methyl palmitate ≥97%
Methyl palmitate

Purity: 97% CAS No.: 112-39-0 Molecular Formula: C17H34O2 Formula Weight: 270.46 Flash Point: 110°(230°F) Refractive Index: 1.4512 Melting Point: 28-34° Boiling Point: 135-137°/0.2mm Beilstein Reference: 1780973 MDL No.: MFCD00008994

Vzorec: C₁₇H₃₄O₂
MW: 270,46 g/mol
Bod varu: 135…137 °C (0,2 mmHg)
Bod tání: 28…34 °C
Hustota: 0,852
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008994
CAS číslo: 112-39-0
EINECS: 203-966-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR