Tisk…

Ethyl palmitate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hexadecanoic acid ethyl ester, Hexadecylic acid ethyl ester, Ethyl hexadecanoate, Palmitic acid ethyl ester

M-Clarity™ quality level = MQ200

P9009-5G P9009-25G
SIALP9009-5GEA 2920 CZK
SIALP9009-5G SIALP9009-25G
Ethyl palmitate, Sigma-Aldrich®
Ethyl palmitate
Vzorec: C₁₈H₃₆O₂
MW: 284,48 g/mol
Bod varu: 342 °C (1013 hPa)
Bod tání: 24 °C
Hustota: 0,86 g/cm³ (20 °C)
MDL Number: MFCD00008996
CAS číslo: 628-97-7
EINECS: 211-064-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR