Isopropyl palmitate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Hexadecylic acid isopropyl ester, Palmitic acid isopropyl ester, Hexadecanoic acid isopropyl ester, Isopropyl Palmitate, Isopropyl hexadecanoate
M-Clarity™ quality level = MQ300
W515604-1KG
W515604-9KG
W515604-SAMPLE
SIALW515604-1KGEA
3160
CZK
SIALW515604-1KG
SIALW515604-9KG
SIALW515604-SAMPLE
Isopropyl palmitate, Sigma-Aldrich®
Isopropyl palmitate
Vzorec:
C₁₉H₃₈O₂ MW: 298,51 g/mol Bod varu: 160 °C (2 mmHg) Bod tání: 13,5 °C Hustota: 0,855 g/cm³ (20 °C) |
MDL Number:
MFCD00008993 CAS číslo: 142-91-6 EINECS: 205-571-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...