Methyl enantate for synthesis, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Methyl Heptanoate, Methyl Enanthate, Enanthic acid methyl ester, Heptanoic acid methyll ester, Heptylic acid methyl ester, Heptanoic acid methyl ester, Enanthic acid methyll ester, Methyl heptanoate
M-Clarity™ quality level = MQ200
8.14926.0100
8.14926.0500
8.14926.0100EA
1630
CZK
8.14926.0100
8.14926.0500
Methyl enantate for synthesis, Sigma-Aldrich®
Methyl enantate
Vzorec:
C₈H₁₆O₂ MW: 144,21 g/mol Bod varu: 172 °C (1013 hPa) Bod tání: –56 °C Hustota: 0,88 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009537 CAS číslo: 106-73-0 EINECS: 203-428-8 UN: 3272 ADR: 3,III Merck Index: 12,04695 |
Výsledky specifikačního testu
Solubility | (20 °C) insoluble |
Melting Point | -56 °C |
Molar Mass | 144.21 g/mol |
Boiling Point | 173 °C (1013 hPa) |
Flash Point | 57 °C |
Density | 0.880 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...