Ethyl enantate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
112364-100ML
112364-500ML
SIAL112364-100MLEA
1330
CZK
SIAL112364-100ML
SIAL112364-500ML
Ethyl enantate, Sigma-Aldrich®
Ethyl enantate
Vzorec:
C₉H₁₈O₂ Bod varu: 372 °C (1013 hPa) Bod tání: –66 °C Hustota: 0,868 g/cm³ (20 °C) |
MDL Number:
MFCD00009538 CAS číslo: 106-30-9 EINECS: 203-382-9 Merck Index: 13,03867 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...