Tisk…

2-Ethylhexyl acetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W514705-1KG-K W514705-9KG-K
SIALW514705-1KG-KEA 2850 CZK
SIALW514705-1KG-K SIALW514705-9KG-K
2-Ethylhexyl acetate, Sigma-Aldrich®
2-Ethylhexyl acetate
Vzorec: C₁₀H₂₀O₂
MW: 172,27 g/mol
Bod varu: 198 °C (1013 hPa)
Bod tání: –45 °C
Hustota: 0,8726 g/cm³ (20 °C)
MDL Number: MFCD00027249
CAS číslo: 103-09-3
EINECS: 203-079-1
Merck Index: 12,06860

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR