Propyl butyrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W293407-1KG-K
W293407-4KG-K
W293407-9KG-K
SIALW293407-1KG-KEA
2230
CZK
SIALW293407-1KG-K
SIALW293407-4KG-K
SIALW293407-9KG-K
Propyl butyrate, Sigma-Aldrich®
Propyl butyrate
Vzorec:
C₇H₁₄O₂ |
MDL Number:
MFCD00009396 CAS číslo: 105-66-8 EINECS: 203-320-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...