Tisk…

Isobutyl butyrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W218715-100G-K W218715-1KG-K
SIALW218715-100G-KEA 2800 CZK
SIALW218715-100G-K SIALW218715-1KG-K
Isobutyl butyrate, Sigma-Aldrich®
Isobutyl butyrate
Vzorec: C₈H₁₆O₂
MDL Number: MFCD00027134
CAS číslo: 539-90-2
EINECS: 208-729-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR