Tisk…

Geranyl butyrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Butyric acid geranyl ester, 3,7-Dimethylocta-2,6-dien-1-yl butyrate, Butanoic acid geranyl ester

M-Clarity™ quality level = MQ300

W251216-100G-K W251216-1KG-K W251216-4KG-K W251216-SAMPLE
SIALW251216-100G-KEA 6280 CZK
SIALW251216-100G-K SIALW251216-1KG-K SIALW251216-4KG-K SIALW251216-SAMPLE
Geranyl butyrate, Sigma-Aldrich®
Geranyl butyrate
Vzorec: C₁₄H₂₄O₂
MDL Number: MFCD00036538
CAS číslo: 106-29-6
EINECS: 203-381-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR